Isopropenyl acetate 99%
Leverantör: Thermo Scientific
Fara
152500010 152502500
ACRO152500010EA 1700 SEK
ACRO152500010 ACRO152502500
Isopropenyl acetate 99%
Isopropenyl acetate
Formel: C₅H₈O₂
MW: 100,12 g/mol
Kokpunkt: 94 °C (1013 hPa)
Smältpunkt: –93 °C
Densitet: 0,92 g/cm³
Flampunkt: 10 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008709
CAS nummer: 108-22-5
EINECS: 203-562-7
UN: 2403
ADR: 3,II
Merck Index: 13,05223
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4000 to 1.4020 (20°C, 589 nm)
GC ≥98.5 %