2-Methylnonane ≥98%
Leverantör: Thermo Scientific
Fara
B20849.03 B20849.06 B20849.14
B20849.03EA 985 SEK
B20849.03 B20849.06 B20849.14
2-Methylnonane ≥98%
2-Methylnonane
Purity: 98+% CAS No.: 871-83-0 Molecular Formula: C10H22 Formula Weight: 142.29 Flash Point: 46°(115°F) Refractive Index: 1.410 Odour: Petroleum-like Boiling Point: 166° Beilstein Reference: 1696990 UN No.: 3295 MDL No.: MFCD00027321
Formel: C₁₀H₂₂
MW: 142,28 g/mol
Kokpunkt: 166 °C
Smältpunkt: –74,6 °C
Densitet: 0,72
Flampunkt: 46 °C (115 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00027321
CAS nummer: 871-83-0
EINECS: 212-814-5
UN: 3295
ADR: 3,III