tert-Butylamin ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
2-Aminoisobutane
B0709.0025
B0709-25ML
B0709-500ML
B0709-100ML
TCIAB0709.0025EA
0
SEK
TCIAB0709.0025
TCIAB0709-25ML
TCIAB0709-500ML
TCIAB0709-100ML
tert-Butylamin ≥98.0% (by GC, titration analysis)
tert-Butylamin
Formel:
C₄H₁₁N MW: 73,14 g/mol Kokpunkt: 46°C Smältpunkt: –72 °C Densitet: 0,6965 g/cm³ (20 °C) Flampunkt: –7°C Storage Temperature: Omgivande |
MDL Number:
MFCD00008050 CAS nummer: 75-64-9 EINECS: 200-888-1 UN: 3286 ADR: 3,II Merck Index: 14,01545 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...