Isopentylamine 99%
Leverantör: Thermo Fisher Scientific
Fara
155470250 155470050 155471000
ACRO155470250EA 705 SEK
ACRO155470250 ACRO155470050 ACRO155471000
Isopentylamine 99%
Isopentylamine
Formel: C₅H₁₃N
Kokpunkt: 95…97 °C (760 mmHg)
Smältpunkt: –60 °C
Densitet: 0,75 g/cm³
Flampunkt: 4 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008203
CAS nummer: 107-85-7
EINECS: 203-526-0
UN: 1106
ADR: 3,II
Merck Index: 13,05128
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4100 (20°C, 589 nm)
Water ≤0.5 %
GC ≥98.0 %