1-Chloro-2,2-dimethylpropane ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: Chloro-1-diméthyl-2,2-propane, neo-Pentylchloride
L02365.06 L02365.14
L02365.06EA 1350 SEK
L02365.06 L02365.14
1-Chloro-2,2-dimethylpropane ≥98%
1-Chloro-2,2-dimethylpropane
Purity: 98% CAS No.: 753-89-9 Molecular Formula: C5H11Cl Formula Weight: 106.60 Flash Point: -8°(17°F) Refractive Index: 1.4039 Odour: Sweet Melting Point: -20° Boiling Point: 83-84° Beilstein Reference: 1730985 UN No.: 1107 MDL No.: MFCD00000941
Formel: C₅H₁₁Cl
MW: 106,6 g/mol
Kokpunkt: 83…84 °C
Smältpunkt: –20 °C
Densitet: 0,868
Flampunkt: –8 °C (17 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000941
CAS nummer: 753-89-9
EINECS: 212-040-8
UN: 1107
ADR: 3,II