3-Chloroacetylacetone 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Chloro-2,4-pentanedione, 3-Chloropentane-2,4-dione
210670050 210671000
ACRO210670050EA 515 SEK
ACRO210670050 ACRO210671000
3-Chloroacetylacetone 98%
3-Chloroacetylacetone
Formel: C₅H₇ClO₂
MW: 134,56 g/mol
Kokpunkt: 49…52 °C (18 mmHg)
Smältpunkt: –15 °C
Densitet: 1,179 g/cm³
Flampunkt: 56 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00009651
CAS nummer: 1694-29-7
EINECS: 216-902-4
UN: 1224
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4825 to 1.4845 (20°C, 589 nm)
GC ≥97.5 %