2,6-Dimethyl-3,5-heptanedione 99%
Leverantör: Thermo Scientific
Varning
408360050 408360250
ACRO408360050EA 2400 SEK
ACRO408360050 ACRO408360250
2,6-Dimethyl-3,5-heptanedione 99%
2,6-Dimethyl-3,5-heptanedione
Formel: C₉H₁₆O₂
MW: 156,22 g/mol
Kokpunkt: 66 °C (8 mmHg)
Densitet: 1,4586 g/cm³
Flampunkt: 60 °C
MDL Number: MFCD00015040
CAS nummer: 18362-64-6
EINECS: 242-234-8
UN: 1224
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4560 to 1.4600 (20°C, 589 nm)
GC ≥98.5 %