3-Metylbutanon ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: Isopropyl Methyl Ketone, 3-Methyl-2-butanone
B24527.AK B24527.AU
B24527.AKEA 1075 SEK
B24527.AK B24527.AU
3-Metylbutanon ≥98%
3-Metylbutanon
Purity: 98% CAS No.: 563-80-4 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: -1°(30°F) Refractive Index: 1.3880 Odour: Fruit-like Melting Point: -92° Boiling Point: 94-95° Beilstein Reference: 1071238 Merck Reference: 14,6090 UN No.: 2397 MDL No.: MFCD00008919
Formel: C₅H₁₀O
MW: 86,13 g/mol
Kokpunkt: 94…95 °C
Smältpunkt: –92 °C
Densitet: 0,805
Flampunkt: –1 °C (30 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008919
CAS nummer: 563-80-4
EINECS: 209-264-3
UN: 2397
ADR: 3,II
Merck Index: 14,6090