2,6-Dimetyl-4-heptanon ≥90%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Diisobutyl Ketone
L16015.AE L16015.AP L16015.0F
L16015.AEEA 585 SEK
L16015.AE L16015.AP L16015.0F
2,6-Dimetyl-4-heptanon ≥90%
2,6-Dimetyl-4-heptanon
Purity: 90% (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone) CAS No.: 108-83-8 Molecular Formula: C9H18O Formula Weight: 142.24 Flash Point: 47°(117°F) Refractive Index: 1.4120 Odour: Unpleasant Melting Point: -42° Boiling Point: 163-173° Beilstein Reference: 1743163 UN No.: 1224 MDL No.: MFCD00008940
Formel: C₉H₁₈O
Kokpunkt: 163…173 °C
Smältpunkt: –42 °C
Densitet: 0,809
Flampunkt: 47 °C (117 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008940
CAS nummer: 108-83-8
EINECS: 203-620-1
UN: 1224
ADR: 3,III