1-Bromo-3-metylbutan 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Methylbutylbromide
403051000 403055000 403050050
ACRO403051000EA 470 SEK
ACRO403051000 ACRO403055000 ACRO403050050
1-Bromo-3-metylbutan 99%
1-Bromo-3-metylbutan
Formel: C₅H₁₁Br
MW: 151,05 g/mol
Kokpunkt: 119 °C (1013 hPa)
Smältpunkt: –112 °C
Densitet: 1,2 g/cm³
Flampunkt: 32 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000253
CAS nummer: 107-82-4
EINECS: 203-522-9
UN: 2341
ADR: 3,III
Merck Index: 13,05130
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4399 to 1.4419 (20°C, 589 nm)
Water ≤0.05 % (K.F.)
GC ≥98.5 %
Acidity ≤0.1 % (as HBr)