n-Pentadekan 99
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: N-Pentadecane, Pentadecane
A10336.14 A10336.18 A10336.30 A10336.36
A10336.14EA 730 SEK
A10336.14 A10336.18 A10336.30 A10336.36
n-Pentadekan 99
n-Pentadekan
Purity: 99% CAS No.: 629-62-9 Molecular Formula: CH3(CH2)13CH3 Formula Weight: 212.42 Flash Point: 132°(269°F) Refractive Index: 1.4320 Melting Point: 9-10° Boiling Point: 269-270° Beilstein Reference: 1698194 MDL No.: MFCD00008990
Formel: H₃C(CH₂)₁₃CH₃
MW: 212.42 g/mol
Kokpunkt: 269…270 °C
Smältpunkt: 9…10 °C
Densitet: 0.769
Flampunkt: 132 °C (269 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008990
CAS nummer: 629-62-9
EINECS: 211-098-1
UN: 3295
ADR: 3 III