cis-3-Hexene ≥97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: (Z)-3-Hexene
H53485.06 H53485.14
H53485.06EA 2175 SEK
H53485.06 H53485.14
cis-3-Hexene ≥97%
cis-3-Hexene
Purity: 97% CAS No.: 7642-09-3 Molecular Formula: C6H12 Formula Weight: 84.16 Flash Point: -25°(-13°F) Refractive Index: 1.395 Boiling Point: 66-68° Beilstein Reference: 1718858 UN No.: 3295 MDL No.: MFCD00085300
Formel: C₆H₁₂
MW: 84.16 g/mol
Kokpunkt: 66…68 °C
Densitet: 0.681
Flampunkt: –25 °C (–13 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00085300
CAS nummer: 7642-09-3
ADR: 3 II