Potassium(4-methoxyphenyl)trifluoroborate 95%
Leverantör: Thermo Scientific
Varning
Synonyms: (4-Methoxyphenyl)trifluoroboric acid potassium salt, Potassium (4-methoxyphenyl)trifluoroborate, Potassium trifluoro(4-methoxyphenyl)borate
H27384.06 H27384.03
H27384.06EA 4350 SEK
H27384.06 H27384.03
Potassium(4-methoxyphenyl)trifluoroborate 95%
Potassium(4-methoxyphenyl)trifluoroborate
Purity: 95% CAS No.: 192863-36-8 Molecular Formula: C7H7BF3KO Formula Weight: 214.04 Melting Point: 300° MDL No.: MFCD04112717
Formel: C₇H₇BF₃KO
MW: 214,04 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD04112717
CAS nummer: 192863-36-8