1,4-Hexadiene cis- and trans-mixture ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: Hexa-1,4-diene
B20060.06 B20060.14
B20060.06EA 1675 SEK
B20060.06 B20060.14
1,4-Hexadiene cis- and trans-mixture ≥99%
1,4-Hexadiene
Purity: 99% CAS No.: 592-45-0 Molecular Formula: CH3CH=CHCH2CH=CH2 Formula Weight: 82.15 Storage Temperature: 2°C to 8°C Flash Point: -26°(-15°F) Refractive Index: 1.4110 Odour: Characteristic Boiling Point: 64-66° Beilstein Reference: 1731889 UN No.: 2458 MDL No.: MFCD00009298
Formel: C₆H₁₀
MW: 82,15 g/mol
Kokpunkt: 64…66 °C
Densitet: 0,710
Flampunkt: –26 °C (–15 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00009298
CAS nummer: 592-45-0
EINECS: 209-756-8
UN: 2458
ADR: 3,II