4'-Phenoxyacetophenone ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A12392.14 A12392.22 A12392.06
A12392.14EA 2675 SEK
A12392.14 A12392.22 A12392.06
4'-Phenoxyacetophenone ≥98%
4'-Phenoxyacetophenone
Purity: 98+% CAS No.: 5031-78-7 Molecular Formula: C14H12O2 Formula Weight: 212.25 Flash Point: 110°(230°F) Melting Point: 48-52° Boiling Point: 325° Beilstein Reference: 2049295 MDL No.: MFCD00008744
Formel: C₁₄H₁₂O₂
Kokpunkt: 325 °C
Smältpunkt: 48…52 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008744
CAS nummer: 5031-78-7