Ammonium cobalt(II) sulphate hexahydrate ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Ammonium cobaltous sulphate hexahydrate, di-Ammonium cobalt (II) sulphate hexahydrate
A10627.14 A10627.22 A10627.36
A10627.14EA 920 SEK
A10627.14 A10627.22 A10627.36
Ammonium cobalt(II) sulphate hexahydrate ≥98%
Ammonium cobalt(II) sulphate hexahydrate
Purity: 98% CAS No.: 13586-38-4 Molecular Formula: (NH4)2Co(SO4)2.6H2O Formula Weight: 395.21 (287.13anhy) Odour: Odourless Melting Point: ca 120° dec. UN No.: 3077 MDL No.: MFCD00151049
Formel: H₈CoN₂O₈S₂·6H₂O
MW: 395.23 g/mol
Smältpunkt: ∼ 120 °C (dec.)
Densitet: 1.902
Storage Temperature: Omgivande
MDL Number: MFCD00151049
CAS nummer: 13586-38-4
EINECS: 237-043-1
ADR: 9 III