Propagyl amine hydrochloride ≥95%
Leverantör: Thermo Scientific
Fara
Synonyms: Propargylamine hydrochloride, Prop-2-yn-1-amine hydrochloride
43775.03 43775.06
43775.03EA 780 SEK
43775.03 43775.06
Propagyl amine hydrochloride ≥95%
Propagyl amine hydrochloride
Purity: 95% CAS No.: 15430-52-1 Molecular Formula: HC≡CCH2NH2.HCl Formula Weight: 91.54 Physical Form: Powder Sensitivity: Hygroscopic UN No.: 2811 MDL No.: MFCD00012907
Formel: C₃H₆ClN
MW: 91,54 g/mol
Smältpunkt: 179…182 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012907
CAS nummer: 15430-52-1
EINECS: 239-440-5
UN: 2811
ADR: 6.1,II