n-Oktadekan ≥99%

Leverantör: Thermo Scientific
Fara

Synonyms: Octadecane

31954.22 31954.36 31954.A3
31954.22EA 1225 SEK
31954.22 31954.36 31954.A3
n-Oktadekan ≥99%
n-Oktadekan

Purity: 99% CAS No.: 593-45-3 Molecular Formula: CH3(CH2)16CH3 Formula Weight: 254.50 Flash Point: 166°(330°F) Physical Form: Crystalline mass/melt Density: 0.782 Refractive Index: 1.4390 Odour: Odourless Melting Point: 28° Boiling Point: 317° MDL No.: MFCD00009007

Formel: H₃C(CH₂)₁₆CH₃
MW: 254,5 g/mol
Kokpunkt: 317 °C
Smältpunkt: 28 °C
Densitet: 0,782
Flampunkt: 166 °C (330 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009007
CAS nummer: 593-45-3
EINECS: 209-790-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR