n-Hexan, EMPARTA® ACS analytiskt reagens, Supelco®
Leverantör: Merck
Fara
Synonyms: Hexyl hydride, Hexanen, Hexane
M-Clarity™ quality level = MQ200
1.07023.2511 1.07023.6025 1.07023.4000
1.07023.2511EA 2170 SEK
1.07023.2511 1.07023.6025 1.07023.4000
n-Hexan, EMPARTA® ACS analytiskt reagens, Supelco®
n-Hexan
Merck Millipore offers EMPARTA® grade solvents for routine tasks in analytical laboratories. EMPARTA® grade solvents offer fewer test parameters than EMSURE®, yet are still of high quality. EMPARTA® solvents meet ACS requirements, making them ideal for a wide range of analytical applications.
Formel: H₃C(CH₂)₄CH₃
MW: 86,18 g/mol
Kokpunkt: 69 °C (1013 hPa)
Smältpunkt: –94,3 °C
Densitet: 0,659 g/cm³ (20 °C)
Flampunkt: –22 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009520
CAS nummer: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM