n-Hentriacontane, Supelco®
Leverantör: Merck
M-Clarity™ quality level = MQ100
51529-1G
51529-250MG
51529-1GEA
2170
SEK
51529-1G
51529-250MG
n-Hentriacontane, Supelco®
n-Hentriacontane
Formel:
H₃C(CH₂)₂₉CH₃ MW: 436.85 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00048745 CAS nummer: 630-04-6 |