Sodium tridecyl sulphate 99% may cont. up to ca 2% water and residual solvent
Leverantör: Thermo Fisher Scientific
Varning
B22089.03 B22089.06 B22089.14
B22089.03EA 670 SEK
B22089.03 B22089.06 B22089.14
Sodium tridecyl sulphate 99% may cont. up to ca 2% water and residual solvent
Sodium tridecyl sulphate
Purity: 99% CAS No.: 3026-63-9 Molecular Formula: C13H27NaO4S Formula Weight: 302.40 Melting Point: 185-189° MDL No.: MFCD00024993
Formel: C₁₃H₂₇NaO₄S
Smältpunkt: 185…189 °C
Storage Temperature: Omgivande
MDL Number: MFCD00024993
CAS nummer: 3026-63-9
EINECS: 221-188-2