Sodium octyl sulphate ≥99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Octyl sulphate, sodium salt, Octyl sulphuric acid salt
A11786.03 A11786.06 A11786.14
A11786.03EA 1275 SEK
A11786.03 A11786.06 A11786.14
Sodium octyl sulphate ≥99%
Sodium octyl sulphate
Purity: 99% CAS No.: 142-31-4 Molecular Formula: CH3(CH2)7OSO3Na Formula Weight: 232.28 Melting Point: ca 195° dec. Beilstein Reference: 3658635
Formel: C₈H₁₇NaO₄S
MW: 232.28 g/mol
Smältpunkt: ∼195 °C (dec.)
Densitet: 1.266 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007470
CAS nummer: 142-31-4
EINECS: 205-535-5