Natriumdodecylsulfat, Supelco®
Leverantör: Merck
|
Fara
|
Synonyms:
Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
M-Clarity™ quality level = MQ100
71726-10G-F
71726-50G-F
71726-10G-FEA
1310
SEK
71726-10G-F
71726-50G-F
Natriumdodecylsulfat, Supelco®
Natriumdodecylsulfat
Formel:
CH₃(CH₂)₁₁OSO₃Na MW: 288,38 g/mol Smältpunkt: 204…207 °C Densitet: 1,1 g/cm³ (20 °C) Flampunkt: >100 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00036175 CAS nummer: 151-21-3 EINECS: 205-788-1 UN: 1325 ADR: 4.1,II Merck Index: 14,08636 |