Natriumdodecylsulfat
Leverantör: RESEARCH ORGANICS
Fara
Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
M-Clarity™ quality level = MQ300
RES9010L-A103X RES9010L-A102X RES9010L-A104X
RESERES9010L-A103XEA 0 SEK
RESERES9010L-A103X RESERES9010L-A102X RESERES9010L-A104X
Natriumdodecylsulfat
Natriumdodecylsulfat
Formel: CH₃(CH₂)₁₁OSO₃Na
MW: 288,38 g/mol
Smältpunkt: 204…207 °C
Densitet: 1,1 g/cm³ (20 °C)
Flampunkt: >100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00036175
CAS nummer: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM