1,2-Ethanedithiol ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 1.2-Ethanedithiol, 1,2-Dimercaptoethane
L12865.14 L12865.22 L12865.36
L12865.14EA 635 SEK
L12865.14 L12865.22 L12865.36
1,2-Ethanedithiol ≥98%
1,2-Ethanedithiol
Purity: 98+% CAS No.: 540-63-6 Molecular Formula: C2H6S2 Formula Weight: 94.20 Flash Point: 45°(113°F) Refractive Index: 1.5580 Odour: Stench Melting Point: -41° Boiling Point: 144-146° Beilstein Reference: 505946 Merck Reference: 14,3725 UN No.: 3071 MDL No.: MFCD00004892
Formel: C₂H₆S₂
MW: 94,2 g/mol
Kokpunkt: 144…146 °C
Smältpunkt: –41 °C
Densitet: 1,123
Flampunkt: 45 °C (113 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004892
CAS nummer: 540-63-6
EINECS: 208-752-3
UN: 3071
ADR: 6.1,II
Merck Index: 14,3725