1-Octanethiol ≥98%

Leverantör: Thermo Scientific
Varning

Synonyms: n-Octyl mercaptan, 1-Mercaptooctane, n-Octanethiol

L07195.AE L07195.AP
L07195.AEEA 870 SEK
L07195.AE L07195.AP
1-Octanethiol ≥98%
1-Octanethiol

Purity: 98% CAS No.: 111-88-6 Molecular Formula: CH3(CH2)7SH Formula Weight: 146.29 Flash Point: 68°(154°F) Refractive Index: 1.4550 Melting Point: -49° Boiling Point: 197-199° Beilstein Reference: 1733101 UN No.: 3082 MDL No.: MFCD00004912

Formel: C₈H₁₈S
MW: 146,3 g/mol
Kokpunkt: 197…199 °C
Smältpunkt: –49 °C
Densitet: 0,843
Flampunkt: 68 °C (154 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004912
CAS nummer: 111-88-6
EINECS: 203-918-1
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR