Metanol, Prepsolv® for preparative chromatography, Supelco®
Leverantör: Merck
Fara
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
M-Clarity™ quality level = MQ200
1.13351.2500 1.13351.4000 1.13351.9030
1.13351.2500EA 830 SEK
1.13351.2500 1.13351.4000 1.13351.9030
Metanol, Prepsolv® for preparative chromatography, Supelco®
Metanol
Formel: CH₃OH
MW: 32.04 g/mol
Kokpunkt: 64,6
Smältpunkt: –98 °C
Densitet: 0.7918 g/cm³ (20 °C)
Flampunkt: 11 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00004595
CAS nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1)
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM