Metanol, BAKER ANALYZED® for LC-MS, suitable for UPLC/UHPLC instruments, J.T.Baker®
Leverantör: Avantor
Fara
9822.1000GLEA 605 SEK
9822.1000GL 9822.2500GL
Metanol, BAKER ANALYZED® for LC-MS, suitable for UPLC/UHPLC instruments, J.T.Baker®
Metanol
Formel: CH₃OH
Kokpunkt: 64,6 °C (1013 hPa)
Smältpunkt: –98 °C
Densitet: 0,7918 g/cm³ (20 °C)
Flampunkt: 11 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00004595
CAS nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Assay min. 99.8 %
Residue after Evaporation (in ppm) max. 1
Water (H2O) max. 0.02 %
LC-Gradient-Diode Array Detection (AU), test solution is modified with 0.1% (v/v) formic acid:
at 235 nm max. 0.005
at 254 nm max. 0.02
LC-MS Gradient Suitability Test (TIC, 100 to 2000 m/z), test solution is modified with 0.1% (v/v) formic acid:
Positive ESI-MS Sensitive Impurities (as Reserpine) max. 50 ng/ml
Trace Impurities (in ppb)
Aluminium (Al) max. 50
Calcium (Ca) max. 50
Iron (Fe) max. 100
Potassium (K) max. 50
Magnesium (Mg) max. 50
Sodium (Na) max. 50
Ultraviolet Absorbance (1.00-cm path vs water):
at 225 nm max. 0.20
at 280 nm max. 0.01
Element concentrations are at time of lot release.