Metanol ≥99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
L13255.AP L13255.AU L13255.0F
L13255.APEA 595 SEK
L13255.AP L13255.AU L13255.0F
Metanol ≥99%
Metanol
Purity: 99% CAS No.: 67-56-1 Molecular Formula: CH3OH Formula Weight: 32.04 Flash Point: 11°(52°F) Refractive Index: 1.3290 Odour: Mild Melting Point: -98° Boiling Point: 64.7° Beilstein Reference: 1098229 Merck Reference: 14,5957 UN No.: 1230 MDL No.: MFCD00004595
Formel: CH₃OH
MW: 32.04 g/mol
Kokpunkt: 64.7 °C
Smältpunkt: –98 °C
Densitet: 0.791
Flampunkt: 11 °C (52 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004595
CAS nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
REACH: 01-2119433307-44
Merck Index: 14,5957
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM