Metanol ≥99.9% semiconductor grade
Leverantör: Thermo Fisher Scientific
|
Fara
|
Synonyms:
Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
19393.K2
19393.M1
19393.K2EA
1275
SEK
19393.K2
19393.M1
Metanol ≥99.9% semiconductor grade
Metanol
Purity: 99.9% min
CAS No.: 67-56-1
Molecular Formula: CH3OH
Formula Weight: 32.04
Flash Point: 11°(52°F)
Physical Form: Liquid
Density: 0.791
Refractive Index: 1.3290
Odour: Mild
Melting Point: -98°
Boiling Point: 64.7°
Beilstein Reference: 1098229
Merck Reference: 14,5957
UN No.: 1230
MDL No.: MFCD00004595
Formel:
CH₃OH Kokpunkt: 64,7 °C Smältpunkt: –98 °C Densitet: 0,791 Flampunkt: 11 °C (52 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00004595 CAS nummer: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3,II REACH: 01-2119433307-44 Merck Index: 14,5957 |