(±)-2-Decanol 98%
Leverantör: Thermo Scientific
Synonyms: (±)-Methyl octyl carbinol, (±)-2-Hydroxy decane, 2-Decanol, (±)-Octyl methyl carbinol
331600050 331600250
ACRO331600050EA 465 SEK
ACRO331600050 ACRO331600250
(±)-2-Decanol 98%
(±)-2-Decanol
Formel: C₁₀H₂₂O
MW: 158,28 g/mol
Kokpunkt: 211 °C (1013 hPa)
Smältpunkt: –5 °C
Densitet: 0,82 g/cm³
Flampunkt: 85 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004594
CAS nummer: 1120-06-5
EINECS: 214-296-6
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4350 (20°C, 589 nm)
GC ≥97.5 %