1-Undecanol 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Undecyl alcohol, Undecyl alcohol, Undecan-1-ol, Decyl carbinol, 1-Hydroxy undecane
140681000 140685000
ACRO140681000EA 625 SEK
ACRO140681000 ACRO140685000
1-Undecanol 98%
1-Undecanol
Formel: C₁₁H₂₄O
MW: 172,31 g/mol
Kokpunkt: 146 °C (30 mmHg)
Smältpunkt: 11 °C
Densitet: 0,831 g/cm³
Flampunkt: >112 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00004751
CAS nummer: 112-42-5
EINECS: 203-970-5
UN: 3082
ADR: 9,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4370 to 1.4420 (20°C, 589 nm)
GC ≥97.5 %
Specific gravity (25/25°C) 0.829 to 0.833