1-Propanol, EMPLURA®, Supelco®
Leverantör: Merck
Fara
Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane
M-Clarity™ quality level = MQ200
1.00996.1000 1.00996.2500 1.00996.6025
1.00996.1000EA 874 SEK
1.00996.1000 1.00996.2500 1.00996.6025
1-Propanol, EMPLURA®, Supelco®
1-Propanol
Formel: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Kokpunkt: 97
Smältpunkt: –127 °C
Densitet: 0.8053 g/cm³ (20 °C)
Flampunkt: 15
Storage Temperature: Omgivande
MDL Number: MFCD00002941
CAS nummer: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3 II
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM