1-Octadecanol ≥97%
Leverantör: Thermo Fisher Scientific
Synonyms: Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol
A12020.0E A12020.30 A12020.36
A12020.0EEA 2300 SEK
A12020.0E A12020.30 A12020.36
1-Octadecanol ≥97%
1-Octadecanol
Purity: 97% CAS No.: 112-92-5 Molecular Formula: CH3(CH2)17OH Formula Weight: 270.50 Flash Point: 174°(345°F) Physical Form: Flakes or pellets Melting Point: 56-60° Boiling Point: 210°/15mm Beilstein Reference: 1362907 Merck Reference: 14,8805 MDL No.: MFCD00002823
Formel: H₃C(CH₂)₁₆CH₂OH
MW: 270.5 g/mol
Kokpunkt: 210 °C (15 mmHg)
Smältpunkt: 56…60 °C
Densitet: 0.805…0.815 g/cm³ (60 °C)
Flampunkt: 174 °C (345 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002823
CAS nummer: 112-92-5
EINECS: 204-017-6
Merck Index: 12,08960