1-Butanol, EMPROVE® ESSENTIAL NF, extra pure suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Fara
Synonyms: 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol
1.00988.1000 1.00988.2500 1.00988.6025
1.00988.1000EA 2200 SEK
1.00988.1000 1.00988.2500 1.00988.6025
1-Butanol, EMPROVE® ESSENTIAL NF, extra pure suitable for use as excipient, SAFC®
1-Butanol
Formel: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Kokpunkt: 117.6
Smältpunkt: –89 °C
Densitet: 0.812 g/cm³ (20 °C)
Flampunkt: 30
Storage Temperature: Omgivande
MDL Number: MFCD00002964
CAS nummer: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3 III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM