Formaldehyd dimetylacetal 99.5+%
Leverantör: Thermo Scientific
Fara
Synonyms: Formal, Methylal, Dimethoxymethane
115560010 115560025 115560250
ACRO115560010EA 1175 SEK
ACRO115560010 ACRO115560025 ACRO115560250
Formaldehyd dimetylacetal 99.5+%
Formaldehyd dimetylacetal
Formel: CH₂(OCH₃)₂
MW: 76,1 g/mol
Kokpunkt: 41…42 °C (760 mmHg)
Smältpunkt: –105 °C
Densitet: 0,86 g/cm³
Flampunkt: –18 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008495
CAS nummer: 109-87-5
EINECS: 203-714-2
UN: 1234
ADR: 3,II
Merck Index: 12,06093
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7dc068e2803e769f0f748f5152cbff2cbfe69fc&vid=511f7642b492436dcce6be18181293c4429a5939&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3525 to 1.3545 (20°C, 589 nm)
Water ≤0.03 %
GC ≥99.50 %