Dietyleter stabilised, TECHNICAL, rectified
Leverantör: VWR Chemicals
Fara
Stabilization: Stabilised med BHT/Ionol 4-10 ppm
23807.366EA 1080 SEK
23807.366 23807.468 23807.550
Dietyleter stabilised, TECHNICAL, rectified
Dietyleter
Formel: (CH₃CH₂)₂O
Kokpunkt: 35 °C (1013 hPa)
Smältpunkt: –123 °C
Densitet: 0,7135 g/cm³ (20 °C)
Flampunkt: –40 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011646
CAS nummer: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Identification Passes test