Dietyleter, vattenfri ≥99% stabilised ACS
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Eter
Stabilization: Stabilised med BHT/Ionol
16767.18 16767.36 16767.62
16767.18EA 1025 SEK
16767.18 16767.36 16767.62
Dietyleter, vattenfri ≥99% stabilised ACS
Dietyleter
Purity: 99% min CAS No.: 60-29-7 Molecular Formula: C4H10O Formula Weight: 74.12 Flash Point: -45°(-49°F) Physical Form: Liquid Density: 0.713 Refractive Index: 1.3525 Appearance: Colourless Odour: Sweet Melting Point: -116° Boiling Point: 34.6° Merck Reference: 14,3806 UN No.: 1155 MDL No.: MFCD00011646
Formel: (CH₃CH₂)₂O
MW: 74.12 g/mol
Kokpunkt: 34.6 °C
Smältpunkt: –116 °C
Densitet: 0.713
Flampunkt: –45 °C (–49 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00011646
CAS nummer: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3 I
REACH: 01-2119535785-29
Merck Index: 14,3806
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM