Hexylamine 99%
Leverantör: Thermo Scientific
Fara
Synonyms: 1-Hexylamine, 1-Aminohexane
204720050 204725000 204721000 204720010
ACRO204720050EA 435 SEK
ACRO204720050 ACRO204725000 ACRO204721000 ACRO204720010
Hexylamine 99%
Hexylamine
Formel: H₃C(CH₂)₄CH₂NH₂
MW: 101,19 g/mol
Kokpunkt: 131 °C (1013 hPa)
Smältpunkt: –19 °C
Densitet: 0,75 g/cm³
Flampunkt: 27 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008240
CAS nummer: 111-26-2
EINECS: 203-851-8
UN: 2734
ADR: 8,I
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4165 to 1.4195 (20°C, 589 nm)
GC ≥98.5 %