Decylamine ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: n-Decylamine, Decan-1-amine, 1-Aminodecane
D0033-25ML D0033-500ML D0033-100ML
TCIAD0033-25MLEA 895 SEK
TCIAD0033-25ML TCIAD0033-500ML TCIAD0033-100ML
Decylamine ≥98.0%
Decylamine
Formel: H₃C(CH₂)₈CH₂NH₂
MW: 157,3 g/mol
Kokpunkt: 220°C
Densitet: 0,787 g/cm³ (20 °C)
Flampunkt: 99°C
Storage Temperature: Omgivande
MDL Number: MFCD00008149
CAS nummer: 2016-57-1
EINECS: 217-957-7
UN: 2922
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acdba1090791775ac2b5dd72fab823ec813d2117&vid=8dfcfbe0de7eaf08c298e5a27d555f242b50cfdc&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %