Tridodecylamine (hydrogen ionophore I) 90%
Leverantör: Thermo Scientific
Varning
Synonyms: Trilaurylamine, Tridodecylamine
421620250 421621000
ACRO421620250EA 1200 SEK
ACRO421620250 ACRO421621000
Tridodecylamine (hydrogen ionophore I) 90%
Tridodecylamine (hydrogen ionophore I)
Formel: C₃₆H₇₅N
MW: 522 g/mol
Kokpunkt: 450 °C (1013 hPa)
Smältpunkt: 16 °C
Densitet: 0,82 g/cm³
Flampunkt: 216 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008971
CAS nummer: 102-87-4
EINECS: 203-063-4
ADR: 9,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4610 (20°C, 589 nm)
Titration with HClO4 ≥88 %