Trietylamin 99%
Leverantör: Thermo Scientific
Fara
Synonyms: (Diethylamino)ethane, TEA
A12646.0F A12646.AK A12646.AP
A12646.0FEA 1950 SEK
A12646.0F A12646.AK A12646.AP
Trietylamin 99%
Trietylamin
Purity: 99% CAS No.: 121-44-8 Molecular Formula: C6H15N Formula Weight: 101.19 Flash Point: -11°(12°F) Refractive Index: 1.4005 Odour: Ammonia-like Melting Point: -115° Boiling Point: 89-90° Beilstein Reference: 1843166 Merck Reference: 14,9666 UN No.: 1296 MDL No.: MFCD00009051
Formel: (C₂H₅)₃N
MW: 101,19 g/mol
Kokpunkt: 89…90 °C
Smältpunkt: –115 °C
Densitet: 0,726
Flampunkt: –11 °C (–12 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009051
CAS nummer: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3,II
Merck Index: 14,9666
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=bed5d56247afa2868328052feb09883049837b93&prid=web&clid=SAPDAM