Etyljodid 98% stabilised, ren
Leverantör: Thermo Scientific
Fara
Synonyms: Ethyl iodide
Stabilization: Stabilised with silver
332121000 332125000 332120025
ACRO332121000EA 1150 SEK
ACRO332121000 ACRO332125000 ACRO332120025
Etyljodid 98% stabilised, ren
Etyljodid
Formel: C₂H₅I
MW: 155,97 g/mol
Kokpunkt: 69…73 °C (760 mmHg)
Smältpunkt: –108 °C
Densitet: 1,95 g/cm³
Flampunkt: 62 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001091
CAS nummer: 75-03-6
EINECS: 200-833-1
ADR: 6.1,III
Merck Index: 13,03846
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow to pink Liquid
Infrared spectrum Conforms
Refractive index 1.5100 to 1.5130 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer silver