1-Iododecane 98% stabilised
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Decane, 1-iodo-, Decyl iodide
330361000 330360250 330365000
ACRO330361000EA 1625 SEK
ACRO330361000 ACRO330360250 ACRO330365000
1-Iododecane 98% stabilised
1-Iododecane
Formel: C₁₀H₂₁I
Kokpunkt: 124 °C (11 mmHg)
Smältpunkt: -16,3 °C
Densitet: 1,25 g/cm³
Flampunkt: 96 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001085
CAS nummer: 2050-77-3
EINECS: 218-104-1
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4840 to 1.4870 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer copper