1,2-Diiodoethane ≥98%
Leverantör: Thermo Fisher Scientific
Varning
L03019.06 L03019.14
L03019.06EA 535 SEK
L03019.06 L03019.14
1,2-Diiodoethane ≥98%
1,2-Diiodoethane
Purity: 98% CAS No.: 624-73-7 Molecular Formula: ICH2CH2I Formula Weight: 281.86 Storage Temperature: 2°C to 8°C Melting Point: 80-84° Beilstein Reference: 1730870 MDL No.: MFCD00001093
Formel: C₂H₄I₂
Kokpunkt: 206 °C (1013 hPa)
Smältpunkt: 80…84 °C
Densitet: 2,130
Storage Temperature: Kylskåp
MDL Number: MFCD00001093
CAS nummer: 624-73-7
EINECS: 210-859-5