Trifluorojodometan ≥97%

Leverantör: Thermo Scientific
Varning

Synonyms: Trifluoromethyl iodide, Iodotrifluoromethane, Perfluoromethyl iodide

41479.14 41479.22
41479.14EA 7400 SEK
41479.14 41479.22
Trifluorojodometan ≥97%
Trifluorojodometan

Purity: 97+% CAS No.: 2314-97-8 Molecular Formula: CF3I Formula Weight: 195.91 Flash Point: None Physical Form: Gas, Packaged in cylinders Density: 2.361 Refractive Index: 1.3790 Sensitivity: Light Sensitive Boiling Point: -22.5° Beilstein Reference: 1732740 UN No.: 1956 MDL No.: MFCD00001060

Formel: CF₃I
MW: 195,91 g/mol
Kokpunkt: –22,5 °C
Smältpunkt: –110 °C
Densitet: 2,361
Storage Temperature: Omgivande
MDL Number: MFCD00001060
CAS nummer: 2314-97-8
EINECS: 219-014-5
UN: 1956
ADR: 2.2,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR