1,1,1-Trifluoroacetylacetone

Fara

Synonyms: 1,1,1-Trifluoropentane-2,4-dione, 1,1,1-Trifluoro-2,4-pentanedione

SB01808DA
MAYBSB01808DAEA 0 SEK
MAYBSB01808DA
1,1,1-Trifluoroacetylacetone
1,1,1-Trifluoroacetylacetone
Formel: C₅H₅F₃O₂
MW: 154,09 g/mol
Kokpunkt: 107 °C (1013 hPa)
Densitet: 1,28 g/cm³ (20 °C)
Flampunkt: 28 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000427
CAS nummer: 367-57-7
EINECS: 206-698-5
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR