6-Chloro-1-hexanol ≥97%
Leverantör: Thermo Fisher Scientific
Varning
A14582.14 A14582.22 A14582.36
A14582.14EA 1400 SEK
A14582.14 A14582.22 A14582.36
6-Chloro-1-hexanol ≥97%
6-Chloro-1-hexanol
Purity: 97% CAS No.: 2009-83-8 Molecular Formula: Cl(CH2)6OH Formula Weight: 136.62 Flash Point: 98°(208°F) Refractive Index: 1.4560 Boiling Point: 109-111°/14mm Beilstein Reference: 1697307 MDL No.: MFCD00002984
Formel: C₆H₁₃ClO
Kokpunkt: 109…111 °C (14 mmHg)
Densitet: 1,027
Flampunkt: 98 °C (208 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002984
CAS nummer: 2009-83-8
EINECS: 217-925-2