2,2,2-Trichloroethanol ≥99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 2,2,2-Trichloroethan-1-ol
L08163.22 L08163.36 L08163.06 L08163.14
L08163.22EA 1100 SEK
L08163.22 L08163.36 L08163.06 L08163.14
2,2,2-Trichloroethanol ≥99%
2,2,2-Trichloroethanol
Purity: 99% CAS No.: 115-20-8 Molecular Formula: Cl3CCH2OH Formula Weight: 149.40 Flash Point: 110°(230°F) Refractive Index: 1.4895 Melting Point: 17-18° Boiling Point: 151-153° Beilstein Reference: 1697495 Merck Reference: 14,9638 MDL No.: MFCD00004677
Formel: C₂H₃Cl₃O
Kokpunkt: 151…153 °C
Smältpunkt: 17…18 °C
Densitet: 1,560
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004677
CAS nummer: 115-20-8
EINECS: 204-071-0
UN: 2810
ADR: 6.1,III
Merck Index: 14,9638