1,3-Diklor-2-propanol 99%
Leverantör: Thermo Scientific
Fara
180180010 180182500
ACRO180180010EA 2275 SEK
ACRO180180010 ACRO180182500
1,3-Diklor-2-propanol 99%
1,3-Diklor-2-propanol
Formel: ClH₂CCHOHCH₂Cl
MW: 128,99 g/mol
Kokpunkt: 174 °C (1013 hPa)
Smältpunkt: –4 °C
Densitet: 1,351 g/cm³
Flampunkt: 74 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000951
CAS nummer: 96-23-1
EINECS: 202-491-9
UN: 2750
ADR: 6.1,II
Merck Index: 13,03100
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4820 to 1.4840 (20°C, 589 nm)
Water ≤0.2 %
GC ≥98.5 %
Acidity (HCl) ≤0.1 %